Product Details

SNP ID
rs140092421
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:77566137 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATTTCTGGAGGACAATCCTGGCA[A/G]TGCCACCGAACACCCTGGATGGGTT
Phenotype
MIM: 608009
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AK5 PubMed Links

Gene Details

Gene
AK5
Gene Name
adenylate kinase 5
There are no transcripts associated with this gene.

Gene
ZZZ3
Gene Name
zinc finger ZZ-type containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308237.1 1473 Silent Mutation CAC,CAT H343H NP_001295166.1
NM_015534.5 1473 Silent Mutation CAC,CAT H837H NP_056349.1
XM_005270725.3 1473 Silent Mutation CAC,CAT H837H XP_005270782.1
XM_005270726.3 1473 Silent Mutation CAC,CAT H837H XP_005270783.1
XM_005270727.3 1473 Silent Mutation CAC,CAT H836H XP_005270784.1
XM_005270729.4 1473 Silent Mutation CAC,CAT H343H XP_005270786.1
XM_017000970.1 1473 Silent Mutation CAC,CAT H836H XP_016856459.1
XM_017000971.1 1473 Intron XP_016856460.1
XM_017000972.1 1473 Silent Mutation CAC,CAT H342H XP_016856461.1

View Full Product Details