Product Details

SNP ID
rs140105842
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:42734928 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTCTGCCCAAGAGCCCCAGGGGT[C/T]AGCACTGACCACTCTGACACAGGCC
Phenotype
MIM: 610036
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CLDN19 PubMed Links

Gene Details

Gene
CLDN19
Gene Name
claudin 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001123395.1 1024 UTR 3 NP_001116867.1
NM_001185117.1 1024 UTR 3 NP_001172046.1
NM_148960.2 1024 UTR 3 NP_683763.2

View Full Product Details