Product Details

SNP ID
rs140106359
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:38012831 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGGCGGCTTTTCGGAAGGCGGCTA[A/C]GTCCCGGCAGCGGGAACACAGAGAG
Phenotype
MIM: 602790 MIM: 609440
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FHL3 PubMed Links

Gene Details

Gene
FHL3
Gene Name
four and a half LIM domains 3
There are no transcripts associated with this gene.

Gene
UTP11
Gene Name
UTP11, small subunit processome component homolog (S. cerevisiae)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016037.3 116 Missense Mutation AAG,ACG K10T NP_057121.2
XM_011541555.2 116 UTR 5 XP_011539857.1

View Full Product Details