Product Details

SNP ID
rs140115755
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:232956750 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTCATGTCTTAACCTGGTTTCAC[A/G]GCACAACAAACATTTAGGAAGAGAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NTPCR PubMed Links

Gene Details

Gene
NTPCR
Gene Name
nucleoside-triphosphatase, cancer-related
There are no transcripts associated with this gene.

View Full Product Details