Product Details

SNP ID
rs140349108
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:28564773 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATGGCATGCTGTATGAATTCTTC[G/T]TCAAAGTCTACCCCTCCTGTCGGGG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TRNAU1AP PubMed Links

Gene Details

Gene
TRNAU1AP
Gene Name
tRNA selenocysteine 1 associated protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017846.4 617 Missense Mutation GTC,TTC V117F NP_060316.1
XM_017001542.1 617 Missense Mutation GTC,TTC V7F XP_016857031.1

View Full Product Details