Product Details
- SNP ID
-
rs140384531
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
2
- Location
-
Chr.1:158699996 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTAGAATTACAGCCACAATACCATC[A/G]TAGGACATGAAGATGAGCATCACAG
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
OR6K2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79973962] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- OR6K2
- Gene Name
- olfactory receptor family 6 subfamily K member 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001005279.1 |
657 |
Silent Mutation |
TAC,TAT |
Y219Y |
NP_001005279.1 |
View Full Product Details