Product Details
- SNP ID
-
rs140391611
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
7
- Location
-
Chr.1:168696586 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAATTCACAGTCGTATTCAGTCATC[C/T]GGCACATTATGAACTTCCACTGGCG
- Phenotype
-
MIM: 125597
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DPT
PubMed Links
Gene Details
- Gene
- DPT
- Gene Name
- dermatopontin
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001937.4 |
599 |
Missense Mutation |
CAG,CGG |
Q190R |
NP_001928.2 |
View Full Product Details