Product Details

SNP ID
rs140472514
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:54065203 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATATCATTATTGAAGCAGAAACA[C/T]ATCAAACAGGTTAAACATAGGCAAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TCEANC2 PubMed Links

Gene Details

Gene
TCEANC2
Gene Name
transcription elongation factor A N-terminal and central domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153035.2 Intron NP_694580.1
XM_017000293.1 Intron XP_016855782.1

View Full Product Details