Product Details

SNP ID
rs140551310
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:31577328 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTGTGCTGCCGCGGCCGTGCCGA[C/T]GACTGTGCCCTGCCCTACCTGGGCG
Phenotype
MIM: 616064
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TINAGL1 PubMed Links

Gene Details

Gene
TINAGL1
Gene Name
tubulointerstitial nephritis antigen like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204414.1 306 Silent Mutation GAC,GAT D60D NP_001191343.1
NM_001204415.1 306 Intron NP_001191344.1
NM_022164.2 306 Silent Mutation GAC,GAT D60D NP_071447.1
XM_005271106.2 306 Silent Mutation GAC,GAT D60D XP_005271163.1
XM_005271107.2 306 Intron XP_005271164.1
XM_011541946.1 306 Intron XP_011540248.1

View Full Product Details