Product Details

SNP ID
rs140579749
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:19597059 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGGTGGGGAACATGTCTGAGTCGG[A/C]GCTCGGCAGGAAGTGGGACCGGTGT
Phenotype
MIM: 616574
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LOC105378614 PubMed Links

Gene Details

Gene
LOC105378614
Gene Name
uncharacterized LOC105378614
There are no transcripts associated with this gene.

Gene
MINOS1
Gene Name
mitochondrial inner membrane organizing system 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001032363.3 83 Missense Mutation GAG,GCG E5A NP_001027535.1
NM_001204082.1 83 Missense Mutation GAG,GCG E5A NP_001191011.1
NM_001204083.1 83 Missense Mutation GAG,GCG E5A NP_001191012.1
Gene
MINOS1-NBL1
Gene Name
MINOS1-NBL1 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204088.1 83 UTR 5 NP_001191017.1
NM_001204089.1 83 UTR 5 NP_001191018.1

View Full Product Details