Product Details

SNP ID
rs140595317
Assay Type
Functionally tested
NCBI dbSNP Submissions
17
Location
Chr.1:172659309 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTTCTTCCCTGTCCAACCTCTGT[A/G]CCCAGAAGGCCTGGTCAAAGGAGGC
Phenotype
MIM: 134638
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FASLG PubMed Links

Gene Details

Gene
FASLG
Gene Name
Fas ligand
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000639.2 302 Silent Mutation GTA,GTG V36V NP_000630.1
NM_001302746.1 302 Silent Mutation GTA,GTG V36V NP_001289675.1

View Full Product Details