Product Details

SNP ID
rs140601192
Assay Type
Functionally tested
NCBI dbSNP Submissions
16
Location
Chr.1:153358311 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACTTGCAAAATGTCGCAGCTGGAA[C/T]GCAACATAGAGACCATCATCAACAC
Phenotype
MIM: 608198 MIM: 123886
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PGLYRP4 PubMed Links

Gene Details

Gene
PGLYRP4
Gene Name
peptidoglycan recognition protein 4
There are no transcripts associated with this gene.

Gene
S100A9
Gene Name
S100 calcium binding protein A9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002965.3 71 Missense Mutation CGC,TGC R10C NP_002956.1

View Full Product Details