Product Details
- SNP ID
-
rs140604430
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
17
- Location
-
Chr.1:248839079 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CACGGAAAGCGACCTGTTCAGGTGC[A/C]GTGTTCCGCATTCGGCGAATAGTTG
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF672
PubMed Links
Gene Details
- Gene
- ZNF672
- Gene Name
- zinc finger protein 672
View Full Product Details