Product Details

SNP ID
rs140624872
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:205569171 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGCTTCGGCCTGTGCATCGCCTT[C/T]CTGGGGCCCACGCTGCTGGACCTGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MFSD4A PubMed Links

Gene Details

Gene
MFSD4A
Gene Name
major facilitator superfamily domain containing 4A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181644.4 188 Silent Mutation TTC,TTT F34F NP_857595.3
XM_011509229.2 188 Silent Mutation TTC,TTT F34F XP_011507531.1

View Full Product Details