Product Details

SNP ID
rs140667727
Assay Type
Functionally Tested
NCBI dbSNP Submissions
8
Location
Chr.1:56495547 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAACCTCTCATGCATGAGACTGCC[G/T]GCTTCTCTGGCTCCTTCTGATTTGC
Phenotype
MIM: 607125
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
PLPP3 PubMed Links

Gene Details

Gene
PLPP3
Gene Name
phospholipid phosphatase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003713.4 2508 UTR 3 NP_003704.3

View Full Product Details