Product Details

SNP ID
rs140675803
Assay Type
Functionally tested
NCBI dbSNP Submissions
16
Location
Chr.1:109669498 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGAATCAGAAAAGGAGCAGATTC[A/G]CGAAGACATTTTGGAGAACCAGTTT
Phenotype
MIM: 138380 MIM: 138333
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GSTM2 PubMed Links

Gene Details

Gene
GSTM2
Gene Name
glutathione S-transferase mu 2 (muscle)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000848.3 381 Missense Mutation CAC,CGC H96R NP_000839.1
NM_001142368.1 381 Missense Mutation CAC,CGC H96R NP_001135840.1
Gene
GSTM4
Gene Name
glutathione S-transferase mu 4
There are no transcripts associated with this gene.

View Full Product Details