Product Details
- SNP ID
-
rs140771210
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
6
- Location
-
Chr.1:204156785 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCCCAGCACAATCTGTCCTCCCAGC[A/G]ATTGGGAATTCCTAAAGGAAAGATC
- Phenotype
-
MIM: 609859
MIM: 179820
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ETNK2
PubMed Links
Gene Details
- Gene
- ETNK2
- Gene Name
- ethanolamine kinase 2
There are no transcripts associated with this gene.
- Gene
- REN
- Gene Name
- renin
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000537.3 |
754 |
Missense Mutation |
TCG,TTG |
S237L |
NP_000528.1 |
View Full Product Details