Product Details

SNP ID
rs140843933
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:173044724 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTTTGAACTTAAACGTGTTGCAC[A/G]CTTTCCTTTTGTAGGATTTCTCTGG
Phenotype
MIM: 603898
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TNFSF18 PubMed Links

Gene Details

Gene
TNFSF18
Gene Name
tumor necrosis factor superfamily member 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005092.3 Intron NP_005083.2

View Full Product Details