Product Details

SNP ID
rs140855313
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:2024975 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGACATCGGCGACTACGTGGGCTC[C/T]AACCTGGAGATCTCCTGGCTCCCCA
Phenotype
MIM: 137163
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GABRD PubMed Links

Gene Details

Gene
GABRD
Gene Name
gamma-aminobutyric acid type A receptor delta subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000815.4 197 Silent Mutation TCC,TCT S34S NP_000806.2
XM_011541194.2 197 Silent Mutation TCC,TCT S47S XP_011539496.1
XM_017000936.1 197 Silent Mutation TCC,TCT S269S XP_016856425.1
Gene
LOC105378589
Gene Name
uncharacterized LOC105378589
There are no transcripts associated with this gene.

View Full Product Details