Product Details

SNP ID
rs140944848
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:63370981 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTCCCTAAATTTGAAGAACTATG[A/G]AGAAATGGTACTTGATGACAGTAGT
Phenotype
MIM: 604566
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ALG6 PubMed Links

Gene Details

Gene
ALG6
Gene Name
ALG6, alpha-1,3-glucosyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013339.3 309 Missense Mutation AAG,GAG K2E NP_037471.2

View Full Product Details