Product Details
- SNP ID
-
rs140964607
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
2
- Location
-
Chr.1:159808339 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGAAAACCAGACTCTGTCCATGGGA[A/G]CAGCAACAGTGCAGAGCCGTGGCCA
- Phenotype
-
MIM: 613562
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FCRL6
PubMed Links
Gene Details
- Gene
- FCRL6
- Gene Name
- Fc receptor like 6
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001004310.2 |
332 |
Missense Mutation |
ACA,GCA |
T72A |
NP_001004310.2 |
NM_001284217.1 |
332 |
Missense Mutation |
ACA,GCA |
T79A |
NP_001271146.1 |
XM_005245128.3 |
332 |
Missense Mutation |
ACA,GCA |
T82A |
XP_005245185.1 |
XM_005245129.3 |
332 |
Missense Mutation |
ACA,GCA |
T67A |
XP_005245186.1 |
XM_005245131.3 |
332 |
Missense Mutation |
ACA,GCA |
T82A |
XP_005245188.1 |
XM_006711292.3 |
332 |
Missense Mutation |
ACA,GCA |
T82A |
XP_006711355.1 |
XM_011509480.1 |
332 |
Missense Mutation |
ACA,GCA |
T79A |
XP_011507782.1 |
XM_011509481.2 |
332 |
Missense Mutation |
ACA,GCA |
T82A |
XP_011507783.1 |
XM_017001176.1 |
332 |
Missense Mutation |
ACA,GCA |
T35A |
XP_016856665.1 |
XM_017001177.1 |
332 |
Missense Mutation |
ACA,GCA |
T3A |
XP_016856666.1 |
View Full Product Details