Product Details

SNP ID
rs140994188
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:153681238 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATCCCGAGTACTTGGAATTCCTGA[A/G]GCAGTTAAAACACCTGGCCTATGAG
Phenotype
MIM: 108960
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR8083 PubMed Links

Gene Details

Gene
MIR8083
Gene Name
microRNA 8083
There are no transcripts associated with this gene.

Gene
NPR1
Gene Name
natriuretic peptide receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000906.3 1472 Missense Mutation AAG,AGG K327R NP_000897.3
XM_005245218.1 1472 Missense Mutation AAG,AGG K327R XP_005245275.1
XM_017001374.1 1472 Missense Mutation AAG,AGG K327R XP_016856863.1

View Full Product Details