Product Details

SNP ID
rs141030117
Assay Type
Functionally tested
NCBI dbSNP Submissions
19
Location
Chr.1:192809076 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTCCAGCGGGAGAACGATAATGC[A/G]AAGTGCTATGTTCTTGGCTGTTCAA
Phenotype
MIM: 600861
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RGS2 PubMed Links

Gene Details

Gene
RGS2
Gene Name
regulator of G-protein signaling 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002923.3 38 Missense Mutation CAA,CGA Q2R NP_002914.1

View Full Product Details