Product Details

SNP ID
rs141031469
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:158466195 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTATTTGCCTTGGTCATTCCTCTG[C/T]TACTTATCCTAGTCTCCTACATCCG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR10K1 PubMed Links

Gene Details

Gene
OR10K1
Gene Name
olfactory receptor family 10 subfamily K member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004473.1 634 Silent Mutation CTA,TTA L212L NP_001004473.1

View Full Product Details