Product Details

SNP ID
rs141067273
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:17413178 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGGCCCCCCAGAAAAACAGACCA[A/C]CTAAGGGAAGACAAAACATGTTCCC
Phenotype
MIM: 609587
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
RCC2 PubMed Links

Gene Details

Gene
RCC2
Gene Name
regulator of chromosome condensation 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136204.2 Intron NP_001129676.1
NM_018715.3 Intron NP_061185.1

View Full Product Details