Product Details

SNP ID
rs141088082
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:247672555 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGTGTTTGGCCCACAGAAAGTCA[A/C]CCTCATGATAAGAGCTGTGTGCACC
Phenotype
MIM: 611677
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
OR13G1 PubMed Links

Gene Details

Gene
OR13G1
Gene Name
olfactory receptor family 13 subfamily G member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005487.1 487 Missense Mutation GTG,TTG V163L NP_001005487.1

View Full Product Details