Product Details
- SNP ID
-
rs141115135
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
15
- Location
-
Chr.1:175157402 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTCAGACCTATGACTCTACCATGG[C/T]GAAGGCGGTGACCAGGGAGGGCTGG
- Phenotype
-
MIM: 616696
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KIAA0040
PubMed Links
Gene Details
- Gene
- KIAA0040
- Gene Name
- KIAA0040
- Gene
- TNN
- Gene Name
- tenascin N
There are no transcripts associated with this gene.
View Full Product Details