Product Details

SNP ID
rs141172283
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:31910064 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTTATAAACTGAACTGCATCTTC[A/G]TACTTCATTCCACATTCAATCAAAG
Phenotype
MIM: 601584
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PTP4A2 PubMed Links

Gene Details

Gene
PTP4A2
Gene Name
protein tyrosine phosphatase type IVA, member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195100.1 Intron NP_001182029.1
NM_001195101.1 Intron NP_001182030.1
NM_080391.3 Intron NP_536316.1
XM_005271231.3 Intron XP_005271288.1
XM_005271232.3 Intron XP_005271289.1
XM_017002401.1 Intron XP_016857890.1

View Full Product Details