Product Details

SNP ID
rs141200690
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:37536866 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAACAACACAACCTGCTTTAGATCT[A/C]TACATGTTATTAGAATAAAGAAAGA
Phenotype
MIM: 608241
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SNIP1 PubMed Links

Gene Details

Gene
SNIP1
Gene Name
Smad nuclear interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024700.3 2188 UTR 3 NP_078976.2

View Full Product Details