Product Details

SNP ID
rs141235412
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:93869919 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCATTTGCTGCTTTTTCAGCCATGA[G/T]CTCTGAGTACTTCCTTCGGTTGTAT
Phenotype
MIM: 611199
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DNTTIP2 PubMed Links

Gene Details

Gene
DNTTIP2
Gene Name
deoxynucleotidyltransferase terminal interacting protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014597.4 2261 Missense Mutation ATC,CTC I735L NP_055412.2

View Full Product Details