Product Details

SNP ID
rs141260822
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:52687794 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTGGGCTCGATTTTTTAGCACCT[A/C]GGCCTTGGCCTCATCCTTATCAACA
Phenotype
MIM: 615623
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
COA7 PubMed Links

Gene Details

Gene
COA7
Gene Name
cytochrome c oxidase assembly factor 7 (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_023077.2 662 Nonsense Mutation GAG,TAG E208* NP_075565.2

View Full Product Details