Product Details
- SNP ID
-
rs141269245
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
6
- Location
-
Chr.1:46402194 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCCCCTGAGGCCGTGCTCTTCACCT[A/G]TGTGGGAAAGGTAAGGCCAGCCAAG
- Phenotype
-
MIM: 602935
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
FAAH
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs566888785] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FAAH
- Gene Name
- fatty acid amide hydrolase
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001441.2 |
381 |
Missense Mutation |
TAT,TGT |
Y100C |
NP_001432.2 |
View Full Product Details