Product Details

SNP ID
rs141368992
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:202012344 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACCCTTCCTTCCTTCCTTGTCAG[C/T]TTCCAGCTCTTCTGATGAGCTCAGT
Phenotype
MIM: 602191 MIM: 602675
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ELF3 PubMed Links

Gene Details

Gene
ELF3
Gene Name
E74 like ETS transcription factor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114309.1 539 Missense Mutation ACT,ATT T129I NP_001107781.1
NM_004433.4 539 Missense Mutation ACT,ATT T129I NP_004424.3
XM_005244942.2 539 Missense Mutation ACT,ATT T129I XP_005244999.1
Gene
MIR6740
Gene Name
microRNA 6740
There are no transcripts associated with this gene.

Gene
RNPEP
Gene Name
arginyl aminopeptidase
There are no transcripts associated with this gene.

View Full Product Details