Product Details

SNP ID
rs141386841
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:40267840 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCTGGAGACTTTCTGGACGGTTC[G/T]GTGGTTATGCTGGCTTTGGACCAGA
Phenotype
MIM: 606480
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZMPSTE24 PubMed Links

Gene Details

Gene
ZMPSTE24
Gene Name
zinc metallopeptidase STE24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005857.4 547 Missense Mutation GGT,TGT G109C NP_005848.2

View Full Product Details