Product Details
- SNP ID
-
rs141467677
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
4
- Location
-
Chr.1:152155206 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCTAGACTGGAATCCCTGTCCCTG[A/G]TTTTGGTACCCTTCCTGTCCTGGAG
- Phenotype
-
MIM: 613259
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
RPTN
PubMed Links
Gene Details
- Gene
- RPTN
- Gene Name
- repetin
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001122965.1 |
1958 |
Silent Mutation |
AAC,AAT |
N631N |
NP_001116437.1 |
View Full Product Details