Product Details

SNP ID
rs141484372
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:86424261 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCTCTACAACATGACCCAAAGGA[A/G]CATTGCAGGTCCTATTTGCAACCTG
Phenotype
MIM: 604003
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLCA2 PubMed Links

Gene Details

Gene
CLCA2
Gene Name
chloride channel accessory 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006536.5 176 Missense Mutation AAC,AGC N5S NP_006527.1
XM_011542448.2 176 Missense Mutation AAC,AGC N5S XP_011540750.1

View Full Product Details