Product Details

SNP ID
rs141562335
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:45500374 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAGCTGAAGCAGAAGATCGAGGA[C/T]ACGCTATGTCCTTTTGGCTTCGAGG
Phenotype
MIM: 609831 MIM: 604746
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC163P PubMed Links

Gene Details

Gene
CCDC163P
Gene Name
coiled-coil domain containing 163, pseudogene
There are no transcripts associated with this gene.

Gene
MMACHC
Gene Name
methylmalonic aciduria (cobalamin deficiency) cblC type, with homocystinuria
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015506.2 49 Silent Mutation GAC,GAT D14D NP_056321.2
XM_005270724.4 49 Silent Mutation GAC,GAT D14D XP_005270781.1
XM_011541204.2 49 UTR 5 XP_011539506.1
Gene
TESK2
Gene Name
testis-specific kinase 2
There are no transcripts associated with this gene.

View Full Product Details