Product Details
- SNP ID
-
rs141634546
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
9
- Location
-
Chr.1:203341955 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGACCAGCCCAGCACAGAGGGAGAG[C/T]TGTCAAGTGCTGCTCACAGACAGCC
- Phenotype
-
MIM: 600245
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FMOD
PubMed Links
Gene Details
- Gene
- FMOD
- Gene Name
- fibromodulin
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002023.4 |
1923 |
UTR 3 |
|
|
NP_002014.2 |
View Full Product Details