Product Details

SNP ID
rs141725098
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:36456271 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGGAAGGTGTACTCAATTCCCAG[A/C]CCCCAGCATATCTTCTCAAAGACAT
Phenotype
MIM: 138971 MIM: 611979 MIM: 608854
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CSF3R PubMed Links

Gene Details

Gene
CSF3R
Gene Name
colony stimulating factor 3 receptor
There are no transcripts associated with this gene.

Gene
MRPS15
Gene Name
mitochondrial ribosomal protein S15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031280.3 716 Silent Mutation GGG,GGT G184G NP_112570.2
Gene
OSCP1
Gene Name
organic solute carrier partner 1
There are no transcripts associated with this gene.

View Full Product Details