Product Details

SNP ID
rs141765942
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:197159023 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAGGAAGAAGAAGGAGAGGAATGA[A/G]ACTAAAGAAGAATAAAAATTTCCAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZBTB41 PubMed Links

Gene Details

Gene
ZBTB41
Gene Name
zinc finger and BTB domain containing 41
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_194314.2 3135 UTR 3 NP_919290.2
XM_011509507.2 3135 Intron XP_011507809.1
XM_017001205.1 3135 Intron XP_016856694.1

View Full Product Details