Product Details

SNP ID
rs141788938
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:16232253 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCTCCCAGGCCTGCCGGAAGGCT[A/G]TGAGGTCCCGCTCGGGCACGTCCGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANO7P1 PubMed Links

Gene Details

Gene
ANO7P1
Gene Name
anoctamin 7 pseudogene 1
There are no transcripts associated with this gene.

Gene
RSG1
Gene Name
REM2 and RAB like small GTPase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030907.3 994 Missense Mutation ACA,ATA T191I NP_112169.2
XM_011542126.2 994 Nonsense Mutation CAG,TAG Q195* XP_011540428.1

View Full Product Details