Product Details

SNP ID
rs141893273
Assay Type
Functionally tested
NCBI dbSNP Submissions
18
Location
Chr.1:152759907 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCATCCCAATCTCAAACTTCCTCT[A/G]TTCAAAGCCAGGCTCCATGCCAATC
Phenotype
MIM: 613260
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KPRP PubMed Links

Gene Details

Gene
KPRP
Gene Name
keratinocyte proline rich protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001025231.1 418 Missense Mutation ATT,GTT I107V NP_001020402.1
XM_011509571.1 418 Missense Mutation ATT,GTT I107V XP_011507873.1

View Full Product Details