Product Details

SNP ID
rs141894100
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:223394100 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGAGGCACTCAGCCTGCGTGTGC[A/G]CCCAGAAGAGAGACAGCAGCGACCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC185 PubMed Links

Gene Details

Gene
CCDC185
Gene Name
coiled-coil domain containing 185
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152610.2 728 Missense Mutation ACC,GCC T209A NP_689823.2

View Full Product Details