Product Details

SNP ID
rs141952266
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:177172746 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCCTCTGTAAAGAATTGTGTGTG[G/T]GCTCTTTGCAGCCCTAGGAGCAGGG
Phenotype
MIM: 600904
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ASTN1 PubMed Links

Gene Details

Gene
ASTN1
Gene Name
astrotactin 1
There are no transcripts associated with this gene.

Gene
BRINP2
Gene Name
BMP/retinoic acid inducible neural specific 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021165.3 Intron NP_066988.1
XM_005245379.1 Intron XP_005245436.1
XM_011509826.1 Intron XP_011508128.1

View Full Product Details