Product Details

SNP ID
rs141955005
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:32779510 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGGGTCCAGAGGTTCAACCTGGC[A/G]GTTTATCCCTTCTCTGGGAAGACAC
Phenotype
MIM: 603623
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KIAA1522 PubMed Links

Gene Details

Gene
KIAA1522
Gene Name
KIAA1522
There are no transcripts associated with this gene.

Gene
YARS
Gene Name
tyrosyl-tRNA synthetase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003680.3 1135 Missense Mutation CGC,TGC R450C NP_003671.1
XM_011542347.1 1135 Missense Mutation CGC,TGC R240C XP_011540649.1
XM_011542348.1 1135 Missense Mutation CGC,TGC R240C XP_011540650.1
XM_017002651.1 1135 Missense Mutation CGC,TGC R240C XP_016858140.1

View Full Product Details