Product Details

SNP ID
rs142004182
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:247100556 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTATGAATTCTTTCGTGGAGGTGAA[C/G]GGAACTGAGGCGACTGAAGGCTTGA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C1orf229 PubMed Links

Gene Details

Gene
C1orf229
Gene Name
chromosome 1 open reading frame 229
There are no transcripts associated with this gene.

Gene
ZNF669
Gene Name
zinc finger protein 669
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142572.1 1095 Missense Mutation CTT,GTT L319V NP_001136044.1
NM_024804.2 1095 Missense Mutation CTT,GTT L405V NP_079080.2
XM_005273270.2 1095 Missense Mutation CTT,GTT L282V XP_005273327.1
XM_005273271.2 1095 Missense Mutation CTT,GTT L282V XP_005273328.1

View Full Product Details