Product Details

SNP ID
rs142022432
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:173627080 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATACAGTACCTTGTCTTCCTTAAG[A/G]AGTTTCCCTGATCCCTTTTCTGTGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANKRD45 PubMed Links

Gene Details

Gene
ANKRD45
Gene Name
ankyrin repeat domain 45
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198493.2 946 Silent Mutation CTC,CTT L192L NP_940895.1
XM_011509466.2 946 Silent Mutation CTC,CTT L192L XP_011507768.1
XM_011509467.1 946 Silent Mutation CTC,CTT L192L XP_011507769.1
XM_011509468.1 946 Silent Mutation CTC,CTT L192L XP_011507770.1
XM_011509470.1 946 Silent Mutation CTC,CTT L192L XP_011507772.1
XM_011509471.2 946 Intron XP_011507773.1
XM_017001123.1 946 Silent Mutation CTC,CTT L212L XP_016856612.1
XM_017001124.1 946 Intron XP_016856613.1
Gene
LOC730159
Gene Name
uncharacterized LOC730159
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195190.1 946 Intron NP_001182119.1
XM_011509968.2 946 Intron XP_011508270.1

View Full Product Details