Product Details

SNP ID
rs142051244
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:165665096 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAATGAAGCACGTCCCAGCCTGAA[A/G]CTCAGCTACCACTCTATGAGCCCGT
Phenotype
MIM: 602733 MIM: 604564
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ALDH9A1 PubMed Links

Gene Details

Gene
ALDH9A1
Gene Name
aldehyde dehydrogenase 9 family member A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000696.3 1489 Missense Mutation CTT,TTT L462F NP_000687.3
XM_011509294.2 1489 Missense Mutation CTT,TTT L368F XP_011507596.1
Gene
MGST3
Gene Name
microsomal glutathione S-transferase 3
There are no transcripts associated with this gene.

View Full Product Details