Product Details

SNP ID
rs142072279
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:153740682 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAAGGTGTATGAGCATTGTGACAT[C/T]GATGACTGCTGGTGTCTCGGAGAGA
Phenotype
MIM: 611347
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
INTS3 PubMed Links

Gene Details

Gene
INTS3
Gene Name
integrator complex subunit 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001324475.1 220 Missense Mutation TCG,TTG S61L NP_001311404.1
NM_023015.4 220 Missense Mutation TCG,TTG S61L NP_075391.3
XM_005245459.2 220 Missense Mutation TCG,TTG S61L XP_005245516.1
XM_005245461.2 220 Intron XP_005245518.1
XM_006711490.3 220 Missense Mutation TCG,TTG S6L XP_006711553.1
XM_011509906.2 220 Missense Mutation TCG,TTG S6L XP_011508208.1
XM_017002132.1 220 Intron XP_016857621.1

View Full Product Details