Product Details

SNP ID
rs142150013
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:154273489 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGCTTCGGCTTCAGCTTCAGCCC[A/T]GGAGGAGGGATACGTTTCCACGATA
Phenotype
MIM: 605998 MIM: 616472
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
HAX1 PubMed Links

Gene Details

Gene
HAX1
Gene Name
HCLS1 associated protein X-1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001018837.1 Intron NP_001018238.1
NM_006118.3 Intron NP_006109.2
Gene
UBAP2L
Gene Name
ubiquitin associated protein 2 like
There are no transcripts associated with this gene.

View Full Product Details